accessibility menu, dialog, popup

UserName

CH3OOBr

CAS RN 10262-65-4

IUPAC Name: 2-bromo-1-(6-methoxynaphthalen-2-yl)ethan-1-one
Synonyms: 2-bromo-1-(6-methoxynaphthalen-2-yl)ethanone
Molecular Weight (g/mol): 279.13
Molecular Formula: C13H11BrO2
InChi Key: HHHKEQGAGUAOQI-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(C=CC2=C1)C(=O)CBr
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)