missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromoacetyl)-6-methoxynaphthalene, 96%, Thermo Scientific Chemicals

Catalog No. p-4812649
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450060050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450060050 Supplier Thermo Scientific Chemicals Supplier No. 450060050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10262-65-4
Molecular Formula C13H11BrO2
Molecular Weight (g/mol) 279.13
InChI Key HHHKEQGAGUAOQI-UHFFFAOYSA-N
Synonym 2-bromoacetyl-6-methoxynaphthalene, 2-bromo-1-6-methoxy-2-naphthyl ethanone, 2-bromo-6'-methoxy-2'-acetonaphthone, 2-bromo-1-6-methoxynaphthalen-2-yl ethan-1-one, 2-bromo-1-6-methoxynaphthalen-2-yl ethanone, ethanone, 2-bromo-1-6-methoxy-2-naphthalenyl, enamine_005243, 2-bromo-1-6-methoxy-naphthalen-2-yl-ethanone
PubChem CID 193452
IUPAC Name 2-bromo-1-(6-methoxynaphthalen-2-yl)ethanone
SMILES COC1=CC2=C(C=C1)C=C(C=C2)C(=O)CBr

Specifications

Melting Point 112.0°C
Assay Percent Range 95% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 279.13
Percent Purity 96%
Chemical Name or Material 2-(Bromoacetyl)-6-methoxynaphthalene
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only