missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetamide 98.0+%, TCI America™

Supplier:  TCI America B28525G

 View more versions of this product

Catalog No. B28525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Bromoacetamide
89°C
C2H4BrNO
5 g
bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
C(C(=O)N)Br
137.964
137.96
Crystalline Powder
683-57-8
White-Yellow
MFCD00008025
2923
JUIKUQOUMZUFQT-UHFFFAOYSA-N
2-bromoacetamide
69632
≥98.0% (GC,N)

Chemical Identifiers

683-57-8
137.964
JUIKUQOUMZUFQT-UHFFFAOYSA-N
69632
C(C(=O)N)Br
C2H4BrNO
MFCD00008025
bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
2-bromoacetamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AB4587000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions