missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetamide 98.0+%, TCI America™

Catalog No. p-7119805
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28525G 5 g
B285225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28525G Supplier TCI America Supplier No. B28525G
Only null left

Chemical Identifiers

CAS 683-57-8
Molecular Formula C2H4BrNO
Molecular Weight (g/mol) 137.964
MDL Number MFCD00008025
InChI Key JUIKUQOUMZUFQT-UHFFFAOYSA-N
Synonym bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
PubChem CID 69632
IUPAC Name 2-bromoacetamide
SMILES C(C(=O)N)Br

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 5 g
UN Number 2923
Formula Weight 137.96
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoacetamide
RTECSNumber AB4587000
TSCA No