missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-p-xylene 99.0+%, TCI America™

Catalog No. p-7118792
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
B099725G 25 g
B0997500G 500 g
2 options

Catalog No. B099725G

Supplier: TCI America B099725G

Only null left

Chemical Identifiers

CAS 553-94-6
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000074
InChI Key QXISTPDUYKNPLU-UHFFFAOYSA-N
Synonym 2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo
PubChem CID 11121
IUPAC Name 2-bromo-1,4-dimethylbenzene
SMILES CC1=CC=C(C)C(Br)=C1

Specifications

Color Colorless
Boiling Point 201°C
Quantity 25 g
Physical Form Clear Liquid at 20°C
Formula Weight 185.06
Percent Purity ≥99.0% (GC)
Chemical Name or Material 2-Bromo-p-xylene