missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-p-xylene 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 500 g |
| Physical Form | Clear Liquid at 20°C |
| Boiling Point | 201°C |
| Formula Weight | 185.06 |
| Percent Purity | ≥99.0% (GC) |
| Grade | Not Available |
| Chemical Name or Material | 2-Bromo-p-xylene |
Safety and Handling
| TSCA | Yes |