missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-p-xylene, 98+%

CAS: 553-94-6 | C8H9Br | 185.06 g/mol

Supplier:  Thermo Scientific Chemicals A1435822

 View more versions of this product

Catalog No. AAA1435822


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-p-xylene
553-94-6
199°C to 201°C
1.551
MFCD00000074
2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo
CC1=CC=C(C)C(Br)=C1
185.06
185.07
9°C to 10°C
1.347
100 g
C8H9Br
2040350
QXISTPDUYKNPLU-UHFFFAOYSA-N
2-bromo-1,4-dimethylbenzene
11121
≥98%

Chemical Identifiers

553-94-6
185.06
QXISTPDUYKNPLU-UHFFFAOYSA-N
11121
CC1=CC=C(C)C(Br)=C1
C8H9Br
MFCD00000074
2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo
2-bromo-1,4-dimethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 209-055-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only