missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-p-cresol 98.0+%, TCI America™

Catalog No. B080825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B080825G 25 g
B0808500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B080825G Supplier TCI America Supplier No. B080825G

Chemical Identifiers

CAS 6627-55-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00002151
InChI Key MTIDYGLTAOZOGU-UHFFFAOYSA-N
Synonym 2-bromo-p-cresol, phenol, 2-bromo-4-methyl, 3-bromo-4-hydroxytoluene, 2-bromo-4-methyl-phenol, p-cresol, 2-bromo, pubchem4096, acmc-209nul, 4-06-00-02143 beilstein handbook reference, ksc494c6f
PubChem CID 23109
IUPAC Name 2-bromo-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)Br

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-p-cresol
RTECSNumber GO6868200
TSCA Yes