missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-N-(tert-butoxycarbonyl)benzylamine 98.0+%, TCI America™

Catalog No. B49201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49201G 1 g
B49205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49201G Supplier TCI America Supplier No. B49201G
Only null left

Chemical Identifiers

CAS 162356-90-3
Molecular Formula C12H16BrNO2
Molecular Weight (g/mol) 286.169
MDL Number MFCD05664385
InChI Key DFNZFCPEUDSNEO-UHFFFAOYSA-N
Synonym tert-butyl 2-bromobenzylcarbamate, n-boc-2-bromobenzylamine, n-tert-butoxycarbonyl-2-bromobenzylamine, tert-butyl n-2-bromophenyl methyl carbamate, 2-bromo-benzyl-carbamic acid tert-butyl ester, tert-butyl 2-bromobenzyl carbamate, 2-bromobenzyl carbamic acid tert-butyl ester, 1-tert-butoxycarbonylaminomethyl-2-bromobenzene, carbamic acid, 2-bromophenyl methyl-, 1,1-dimethylethyl ester, pubchem7804
PubChem CID 7018300
IUPAC Name tert-butyl N-[(2-bromophenyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=CC=C1Br

Specifications

Melting Point 53°C
Color White
Quantity 1 g
Formula Weight 286.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-N-(tert-butoxycarbonyl)benzylamine