missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-N,N-dimethylaniline 98.0+%, TCI America™

Catalog No. p-7122830
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32435G 5 g
B324325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32435G Supplier TCI America Supplier No. B32435G

Chemical Identifiers

CAS 698-00-0
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00013522
InChI Key ONMSBNJJCUCYED-UHFFFAOYSA-N
Synonym 2-bromo-n,n-dimethylbenzenamine, n,n-dimethyl-2-bromoaniline, o-bromo-n,n-dimethylaniline, o-bromo-n,n'-dimethylaniline, benzenamine, bromo-n,n-dimethyl, pubchem3759, bromo-n,n-dimethylaniline, acmc-1az6p, 2-bromophenyl dimethylamine, 2-dimethylamino bromobenzene
PubChem CID 136527
IUPAC Name 2-bromo-N,N-dimethylaniline
SMILES CN(C)C1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
Formula Weight 200.08
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromo-N,N-dimethylaniline