accessibility menu, dialog, popup

UserName

CAS RN 698-00-0

IUPAC Name: 2-bromo-N,N-dimethylaniline
Synonyms: 2-bromo-N,N-dimethylaniline
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: ONMSBNJJCUCYED-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results