missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-alpha-methylbenzyl alcohol, 99%

Catalog No. AC393640010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393640010 1 g Glass bottle
AC393640050 5 g Glass bottle
AC393640250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC393640010 Supplier Thermo Scientific Chemicals Supplier No. 393640010

CAS: 5411-56-3 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5411-56-3
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00065001
InChI Key DZLZSFZSPIUINR-UHFFFAOYSA-N
Synonym 1-2-bromophenyl ethanol, 2-bromo-alpha-methylbenzyl alcohol, 1-2-bromophenyl ethan-1-ol, 1-2'-bromophenyl-1-hydroxyethane, acmc-209wki, o-bromo-1-phenylethanol, o-bromo-1-phenyl ethanol, acmc-209y6n, 1-2-bro-mophenyl ethanol, 1-2-bromo-phenyl-ethanol
PubChem CID 95455
IUPAC Name 1-(2-bromophenyl)ethanol
SMILES CC(C1=CC=CC=C1Br)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 54°C to 56°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Linear Formula BrC6H4CH(OH)CH3
Quantity 1 g
Formula Weight 201.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromo-α-methylbenzyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-491-3

RUO – Research Use Only