missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9-phenylcarbazole 98.0+%, TCI America™
$71.23 - $211.77
Chemical Identifiers
| CAS | 94994-62-4 |
|---|---|
| Molecular Formula | C18H12BrN |
| Molecular Weight (g/mol) | 322.205 |
| MDL Number | MFCD24387061 |
| InChI Key | SOODLDGRGXOSTA-UHFFFAOYSA-N |
| PubChem CID | 66838044 |
| IUPAC Name | 2-bromo-9-phenylcarbazole |
| SMILES | C1=CC=C(C=C1)N2C3=CC=CC=C3C4=C2C=C(C=C4)Br |
Chemical Identifiers
| 94994-62-4 | |
| 322.205 | |
| SOODLDGRGXOSTA-UHFFFAOYSA-N | |
| 2-bromo-9-phenylcarbazole |
| C18H12BrN | |
| MFCD24387061 | |
| 66838044 | |
| C1=CC=C(C=C1)N2C3=CC=CC=C3C4=C2C=C(C=C4)Br |
Specifications
| 94994-62-4 | |
| White-Yellow | |
| MFCD24387061 | |
| SOODLDGRGXOSTA-UHFFFAOYSA-N | |
| 2-bromo-9-phenylcarbazole | |
| 66838044 | |
| ≥98.0% (GC,N) | |
| 2-Bromo-9-phenylcarbazole |
| 90°C | |
| C18H12BrN | |
| 1 g | |
| C1=CC=C(C=C1)N2C3=CC=CC=C3C4=C2C=C(C=C4)Br | |
| 322.205 | |
| 322.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No