accessibility menu, dialog, popup

UserName

CAS RN 94994-62-4

NBr

CAS RN 94994-62-4

IUPAC Name: 2-bromo-9-phenyl-9H-carbazole
Synonyms: 2-bromo-9-phenylcarbazole
Molecular Weight (g/mol): 322.21
Molecular Formula: C18H12BrN
InChi Key: SOODLDGRGXOSTA-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results