missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9-phenylcarbazole 98.0+%, TCI America™

Catalog No. B44391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44391G 1 g
B44395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44391G Supplier TCI America Supplier No. B44391G
Only null left

Chemical Identifiers

CAS 94994-62-4
Molecular Formula C18H12BrN
Molecular Weight (g/mol) 322.205
MDL Number MFCD24387061
InChI Key SOODLDGRGXOSTA-UHFFFAOYSA-N
PubChem CID 66838044
IUPAC Name 2-bromo-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=CC=CC=C3C4=C2C=C(C=C4)Br

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 1 g
Formula Weight 322.21
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9-phenylcarbazole