missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9-n-octyl-9H-carbazole 98.0+%, TCI America™

Catalog No. B56091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56091G 1 g
B56095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56091G Supplier TCI America Supplier No. B56091G
Only null left

Chemical Identifiers

CAS 1356465-23-0
Molecular Formula C20H24BrN
Molecular Weight (g/mol) 358.32
MDL Number MFCD31618111
InChI Key QLOUQHQDDYJWRM-UHFFFAOYSA-N
PubChem CID 129319318
IUPAC Name 2-bromo-9-octyl-9H-carbazole
SMILES CCCCCCCCN1C2=C(C=CC=C2)C2=C1C=C(Br)C=C2

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 1 g
Formula Weight 358.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9-n-octyl-9H-carbazole