accessibility menu, dialog, popup

UserName

CAS RN 1356465-23-0

H3CNBr

CAS RN 1356465-23-0

IUPAC Name: 2-bromo-9-octyl-9H-carbazole
Synonyms: 2-bromo-9-octylcarbazole
Molecular Weight (g/mol): 358.32
Molecular Formula: C20H24BrN
InChi Key: QLOUQHQDDYJWRM-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C2=C(C=CC=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results