missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9-fluorenone, 96%

Catalog No. p-7037264
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5596203 1 g
AAH5596206 5 g
2 options

Catalog No. AAH5596203

Supplier: Thermo Scientific Chemicals H5596203

Only null left

CAS: 3096-56-8 | C13H7BrO | 259.10 g/mol

2-Bromo-9-fluorenone, is used as organic intermediates functional materials. It is mainly used as best liquid crystal material. It is used as an OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-9-fluorenone, is used as organic intermediates functional materials. It is mainly used as best liquid crystal material. It is used as an OLED intermediate.

Solubility
Solubility in other solvents, soluble in acetone.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 3096-56-8
Molecular Formula C13H7BrO
Molecular Weight (g/mol) 259.10
MDL Number MFCD00010789
InChI Key MTCARZDHUIEYMB-UHFFFAOYSA-N
Synonym 2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f
PubChem CID 725831
IUPAC Name 2-bromofluoren-9-one
SMILES BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 146°C to 148°C
Quantity 1 g
Formula Weight 259.1
Percent Purity 96%
Chemical Name or Material 2-Bromo-9-fluorenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only