missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9,9-diphenylfluorene 98.0+%, TCI America™

$59.65 - $186.41

Chemical Identifiers

CAS 474918-32-6
Molecular Formula C25H17Br
Molecular Weight (g/mol) 397.315
MDL Number MFCD09907957
InChI Key WNXNWOBGPRKOJF-UHFFFAOYSA-N
PubChem CID 46856308
IUPAC Name 2-bromo-9,9-diphenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)Br)C5=CC=CC=C5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B44501G
View Documents
TCI America
B44501G
1 g
Each for $59.65
Only null left
 
B44505G
View Documents
TCI America
B44505G
5 g
Each for $186.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

474918-32-6
397.315
WNXNWOBGPRKOJF-UHFFFAOYSA-N
2-bromo-9,9-diphenylfluorene
C25H17Br
MFCD09907957
46856308
C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)Br)C5=CC=CC=C5

Specifications

474918-32-6
White
MFCD09907957
WNXNWOBGPRKOJF-UHFFFAOYSA-N
2-bromo-9,9-diphenylfluorene
46856308
≥98.0% (GC)
2-Bromo-9,9-diphenylfluorene
219°C
C25H17Br
1 g
C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)Br)C5=CC=CC=C5
397.315
397.32
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'