missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9,9-diphenylfluorene 98.0+%, TCI America™

Catalog No. B44505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44505G 5 g
B44501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44505G Supplier TCI America Supplier No. B44505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 474918-32-6
Molecular Formula C25H17Br
Molecular Weight (g/mol) 397.315
MDL Number MFCD09907957
InChI Key WNXNWOBGPRKOJF-UHFFFAOYSA-N
PubChem CID 46856308
IUPAC Name 2-bromo-9,9-diphenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)Br)C5=CC=CC=C5

Specifications

Melting Point 219°C
Color White
Quantity 5 g
Formula Weight 397.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9,9-diphenylfluorene