missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9,9-diphenylfluorene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 219°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 397.32 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Bromo-9,9-diphenylfluorene |
Safety and Handling
| TSCA | No |