missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9,9-difluoro-7-iodo-9H-fluorene 96.0+%, TCI America™

Catalog No. p-7124042
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
B5327200MG 200 mg
B53271G 1 g
2 options

Catalog No. B5327200MG

Supplier: TCI America B5327200MG

Only null left

Chemical Identifiers

CAS 1499193-60-0
Molecular Formula C13H6BrF2I
Molecular Weight (g/mol) 406.996
InChI Key DKSYLEFLMUQPDJ-UHFFFAOYSA-N
PubChem CID 59611589
IUPAC Name 2-bromo-9,9-difluoro-7-iodofluorene
SMILES C1=CC2=C(C=C1Br)C(C3=C2C=CC(=C3)I)(F)F

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 200 mg
Formula Weight 407.00
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9,9-difluoro-7-iodo-9H-fluorene