accessibility menu, dialog, popup

UserName

CAS RN 1499193-60-0

BrIFF

CAS RN 1499193-60-0

IUPAC Name: 2-bromo-9,9-difluoro-7-iodo-9H-fluorene
Synonyms: 2-bromo-9,9-difluoro-7-iodofluorene
Molecular Weight (g/mol): 407
Molecular Formula: C13H6BrF2I
InChi Key: DKSYLEFLMUQPDJ-UHFFFAOYSA-N
SMILES: FC1(F)C2=C(C=CC(Br)=C2)C2=C1C=C(I)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results