missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9,9-di-p-tolyl-9H-fluorene 98.0+%, TCI America™
$288.94 - $864.92
Chemical Identifiers
| CAS | 474918-33-7 |
|---|---|
| Molecular Formula | C27H21Br |
| Molecular Weight (g/mol) | 425.369 |
| InChI Key | PUGVEXPXLSEEOS-UHFFFAOYSA-N |
| PubChem CID | 46856309 |
| IUPAC Name | 2-bromo-9,9-bis(4-methylphenyl)fluorene |
| SMILES | CC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)Br)C5=CC=C(C=C5)C |
Chemical Identifiers
| 474918-33-7 | |
| 425.369 | |
| 46856309 | |
| CC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)Br)C5=CC=C(C=C5)C |
| C27H21Br | |
| PUGVEXPXLSEEOS-UHFFFAOYSA-N | |
| 2-bromo-9,9-bis(4-methylphenyl)fluorene |
Specifications
| 474918-33-7 | |
| White-Yellow | |
| 1 g | |
| CC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)Br)C5=CC=C(C=C5)C | |
| 425.369 | |
| 425.37 | |
| Crystalline Powder |
| 184°C | |
| C27H21Br | |
| PUGVEXPXLSEEOS-UHFFFAOYSA-N | |
| 2-bromo-9,9-bis(4-methylphenyl)fluorene | |
| 46856309 | |
| ≥98.0% (GC) | |
| 2-Bromo-9,9-di-p-tolyl-9H-fluorene |
Safety and Handling
TSCA : No