missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9,9-di-p-tolyl-9H-fluorene 98.0+%, TCI America™

Catalog No. B56195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56195G 5 g
B56191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56195G Supplier TCI America Supplier No. B56195G
Only null left

Chemical Identifiers

CAS 474918-33-7
Molecular Formula C27H21Br
Molecular Weight (g/mol) 425.369
InChI Key PUGVEXPXLSEEOS-UHFFFAOYSA-N
PubChem CID 46856309
IUPAC Name 2-bromo-9,9-bis(4-methylphenyl)fluorene
SMILES CC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)Br)C5=CC=C(C=C5)C

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 5 g
Formula Weight 425.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9,9-di-p-tolyl-9H-fluorene