missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9-(2-naphthyl)-9H-carbazole 98.0+%, TCI America™
$141.67 - $417.56
Chemical Identifiers
| CAS | 1427316-53-7 |
|---|---|
| Molecular Formula | C22H14BrN |
| Molecular Weight (g/mol) | 372.265 |
| MDL Number | MFCD26127422 |
| InChI Key | PYHCFFLUGHKAPZ-UHFFFAOYSA-N |
| PubChem CID | 71302478 |
| IUPAC Name | 2-bromo-9-naphthalen-2-ylcarbazole |
| SMILES | C1=CC=C2C=C(C=CC2=C1)N3C4=CC=CC=C4C5=C3C=C(C=C5)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5051200MG
|
TCI America
B5051200MG |
200 mg |
Each for $141.67
|
|
|||||
|
B50511G
|
TCI America
B50511G |
1 g |
Each for $417.56
|
|
|||||
Chemical Identifiers
| 1427316-53-7 | |
| 372.265 | |
| PYHCFFLUGHKAPZ-UHFFFAOYSA-N | |
| 2-bromo-9-naphthalen-2-ylcarbazole |
| C22H14BrN | |
| MFCD26127422 | |
| 71302478 | |
| C1=CC=C2C=C(C=CC2=C1)N3C4=CC=CC=C4C5=C3C=C(C=C5)Br |
Specifications
| 1427316-53-7 | |
| White-Yellow | |
| MFCD26127422 | |
| PYHCFFLUGHKAPZ-UHFFFAOYSA-N | |
| 2-bromo-9-naphthalen-2-ylcarbazole | |
| 71302478 | |
| ≥98.0% (GC) | |
| 2-Bromo-9-(2-naphthyl)-9H-carbazole |
| 143°C | |
| C22H14BrN | |
| 200 mg | |
| C1=CC=C2C=C(C=CC2=C1)N3C4=CC=CC=C4C5=C3C=C(C=C5)Br | |
| 372.265 | |
| 372.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No