missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9-(2-naphthyl)-9H-carbazole 98.0+%, TCI America™
Supplier: TCI America B5051200MG
Specifications
| 2-Bromo-9-(2-naphthyl)-9H-carbazole | |
| 143°C | |
| C22H14BrN | |
| 200 mg | |
| C1=CC=C2C=C(C=CC2=C1)N3C4=CC=CC=C4C5=C3C=C(C=C5)Br | |
| 372.265 | |
| 372.27 | |
| Crystalline Powder |
| 1427316-53-7 | |
| White-Yellow | |
| MFCD26127422 | |
| PYHCFFLUGHKAPZ-UHFFFAOYSA-N | |
| 2-bromo-9-naphthalen-2-ylcarbazole | |
| 71302478 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1427316-53-7 | |
| 372.265 | |
| PYHCFFLUGHKAPZ-UHFFFAOYSA-N | |
| 2-bromo-9-naphthalen-2-ylcarbazole |
| C22H14BrN | |
| MFCD26127422 | |
| 71302478 | |
| C1=CC=C2C=C(C=CC2=C1)N3C4=CC=CC=C4C5=C3C=C(C=C5)Br |
Safety and Handling
TSCA : No