missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9,10-di(2-naphthyl)anthracene 98.0+%, TCI America™

Catalog No. p-7123469
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43211G 1 g
B43215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43211G Supplier TCI America Supplier No. B43211G
Only null left

Chemical Identifiers

CAS 474688-76-1
Molecular Formula C34H21Br
Molecular Weight (g/mol) 509.446
MDL Number MFCD11045064
InChI Key NNVPXSAMRFIMLP-UHFFFAOYSA-N
PubChem CID 22247157
IUPAC Name 2-bromo-9,10-dinaphthalen-2-ylanthracene
SMILES C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC(=CC4=C(C5=CC=CC=C53)C6=CC7=CC=CC=C7C=C6)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 509.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9,10-di(2-naphthyl)anthracene