missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-8-iododibenzofuran 98.0+%, TCI America™
$143.54 - $492.11
Chemical Identifiers
| CAS | 916435-41-1 |
|---|---|
| Molecular Formula | C12H6BrIO |
| Molecular Weight (g/mol) | 372.987 |
| InChI Key | YMEVNDQRPHHDRH-UHFFFAOYSA-N |
| PubChem CID | 67335932 |
| IUPAC Name | 2-bromo-8-iododibenzofuran |
| SMILES | C1=CC2=C(C=C1Br)C3=C(O2)C=CC(=C3)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4867200MG
|
TCI America
B4867200MG |
200 mg |
Each for $143.54
|
|
|||||
|
B48671G
|
TCI America
B48671G |
1 g |
Each for $492.11
|
|
|||||
Chemical Identifiers
| 916435-41-1 | |
| 372.987 | |
| 67335932 | |
| C1=CC2=C(C=C1Br)C3=C(O2)C=CC(=C3)I |
| C12H6BrIO | |
| YMEVNDQRPHHDRH-UHFFFAOYSA-N | |
| 2-bromo-8-iododibenzofuran |
Specifications
| 916435-41-1 | |
| White | |
| 200 mg | |
| C1=CC2=C(C=C1Br)C3=C(O2)C=CC(=C3)I | |
| 372.987 | |
| 372.99 | |
| Crystalline Powder |
| 173°C | |
| C12H6BrIO | |
| YMEVNDQRPHHDRH-UHFFFAOYSA-N | |
| 2-bromo-8-iododibenzofuran | |
| 67335932 | |
| ≥98.0% (GC) | |
| 2-Bromo-8-iododibenzofuran |
Safety and Handling
TSCA : No