accessibility menu, dialog, popup

UserName

Explore New Spotlight Deals on Equipment, Instruments, and More

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 916435-41-1

BrOI

CAS RN 916435-41-1

IUPAC Name: 4-bromo-12-iodo-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 4-bromo-12-iodo-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 372.99
Molecular Formula: C12H6BrIO
InChi Key: YMEVNDQRPHHDRH-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OC3=C2C=C(I)C=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.