missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-7-methoxy-9H-carbazole 98.0+%, TCI America™

Catalog No. B55171G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B55171G Supplier TCI America Supplier No. B55171G

Chemical Identifiers

CAS 200878-50-8
Molecular Formula C13H10BrNO
Molecular Weight (g/mol) 276.133
InChI Key AHKGUQOMPCEFCL-UHFFFAOYSA-N
PubChem CID 10707575
IUPAC Name 2-bromo-7-methoxy-9H-carbazole
SMILES COC1=CC2=C(C=C1)C3=C(N2)C=C(C=C3)Br

Specifications

Melting Point 284°C
Quantity 1 g
Formula Weight 276.13
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-7-methoxy-9H-carbazole