accessibility menu, dialog, popup

UserName

CH3ONHBr

CAS RN 200878-50-8

IUPAC Name: 2-bromo-7-methoxy-9H-carbazole
Synonyms: 2-bromo-7-methoxy-9H-carbazole
Molecular Weight (g/mol): 276.13
Molecular Formula: C13H10BrNO
InChi Key: AHKGUQOMPCEFCL-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=C1)C1=C(N2)C=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results