missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-6-(tetrahydropyran-4-yloxy)pyridine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC58210DA

 View more versions of this product

Catalog No. CC58210DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-Bromo-6-(tetrahydropyran-4-yloxy)pyridine
96.5
97%
C10H12BrNO2
1g
VLEKIKTZHYMZEW-UHFFFAOYSA-N
2-bromo-6-(oxan-4-yloxy)pyridine
43811040
97%
892502-16-8
100.0
Amber Glass Bottle
MFCD12198112
2-bromo-6-tetrahydropyran-4-yloxy pyridine, 2-bromo-6-oxan-4-yloxy pyridine, 2-bromo-6-oxan-4-yl oxy pyridine, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy-6-bromopyridine
C1COCCC1OC2=NC(=CC=C2)Br
258.115
258.11

Chemical Identifiers

892502-16-8
258.115
VLEKIKTZHYMZEW-UHFFFAOYSA-N
43811040
C1COCCC1OC2=NC(=CC=C2)Br
C10H12BrNO2
MFCD12198112
2-bromo-6-tetrahydropyran-4-yloxy pyridine, 2-bromo-6-oxan-4-yloxy pyridine, 2-bromo-6-oxan-4-yl oxy pyridine, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy-6-bromopyridine
2-bromo-6-(oxan-4-yloxy)pyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions