missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-nitrotoluene 98.0+%, TCI America™

Catalog No. p-7119327
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18785G 5 g
B187825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18785G Supplier TCI America Supplier No. B18785G
Only null left

Chemical Identifiers

CAS 55289-35-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00009792
InChI Key LYTNSGFSAXWBCA-UHFFFAOYSA-N
Synonym 2-bromo-6-nitrotoluene, 6-bromo-2-nitrotoluene, 3-bromo-2-methylnitrobenzene, 1-bromo-2-methyl-3-nitro-benzene, benzene, 1-bromo-2-methyl-3-nitro, bromo 2--6-nitrotoluene, 2-bromo-6-nitrotoluene,2-nitro-6-bromotoluene, 3-bromo-2-methyl-1-nitrobenzene, 1-brom-2-methyl-3-nitrobenzol, pubchem3823
PubChem CID 123537
IUPAC Name 1-bromo-2-methyl-3-nitrobenzene
SMILES CC1=C(C=CC=C1Br)[N+](=O)[O-]

Specifications

Melting Point 40°C
Color Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Bromo-6-nitrotoluene