missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-nitrotoluene, 98%

Catalog No. AAB2165806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2165806 5 g
AAB2165814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2165806 Supplier Thermo Scientific Chemicals Supplier No. B2165806

CAS: 55289-35-5 | C7H6BrNO2 | 216.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55289-35-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00009792
InChI Key LYTNSGFSAXWBCA-UHFFFAOYSA-N
Synonym 2-bromo-6-nitrotoluene, 6-bromo-2-nitrotoluene, 3-bromo-2-methylnitrobenzene, 1-bromo-2-methyl-3-nitro-benzene, benzene, 1-bromo-2-methyl-3-nitro, bromo 2--6-nitrotoluene, 2-bromo-6-nitrotoluene,2-nitro-6-bromotoluene, 3-bromo-2-methyl-1-nitrobenzene, 1-brom-2-methyl-3-nitrobenzol, pubchem3823
PubChem CID 123537
IUPAC Name 1-bromo-2-methyl-3-nitrobenzene
SMILES CC1=C(C=CC=C1Br)[N+](=O)[O-]

Specifications

Melting Point 37°C to 42°C
Boiling Point 108°C to 110°C (3.3 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2502581
Formula Weight 216.04
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-nitrotoluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 259-566-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only