missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-nitrophenol 98.0+%, TCI America™
$78.93 - $232.95
Chemical Identifiers
| CAS | 13073-25-1 |
|---|---|
| Molecular Formula | C6H4BrNO3 |
| Molecular Weight (g/mol) | 218.01 |
| MDL Number | MFCD02683216 |
| InChI Key | VEJSIOPQKQXJAT-UHFFFAOYSA-N |
| Synonym | 2-bromo-6-nitrophenol, phenol, 2-bromo-6-nitro, 2-bromo-6-nitro-phenol, 6-bromo-2-nitrophenol, 6-nitro-2-bromophenol, 3-bromo-2-hydroxynitrobenzene, pubchem3114, 2-brom-6-nitrophenol, 2-nitro-6-bromophenol, acmc-209bjb |
| PubChem CID | 13545453 |
| IUPAC Name | 2-bromo-6-nitrophenol |
| SMILES | OC1=C(Br)C=CC=C1[N+]([O-])=O |
Chemical Identifiers
| 13073-25-1 | |
| 218.01 | |
| VEJSIOPQKQXJAT-UHFFFAOYSA-N | |
| 13545453 | |
| OC1=C(Br)C=CC=C1[N+]([O-])=O |
| C6H4BrNO3 | |
| MFCD02683216 | |
| 2-bromo-6-nitrophenol, phenol, 2-bromo-6-nitro, 2-bromo-6-nitro-phenol, 6-bromo-2-nitrophenol, 6-nitro-2-bromophenol, 3-bromo-2-hydroxynitrobenzene, pubchem3114, 2-brom-6-nitrophenol, 2-nitro-6-bromophenol, acmc-209bjb | |
| 2-bromo-6-nitrophenol |
Specifications
| 13073-25-1 | |
| C6H4BrNO3 | |
| 5 g | |
| VEJSIOPQKQXJAT-UHFFFAOYSA-N | |
| 2-bromo-6-nitrophenol | |
| 13545453 | |
| ≥98.0% (GC,T) | |
| 2-Bromo-6-nitrophenol |
| 68°C | |
| MFCD02683216 | |
| 2-bromo-6-nitrophenol, phenol, 2-bromo-6-nitro, 2-bromo-6-nitro-phenol, 6-bromo-2-nitrophenol, 6-nitro-2-bromophenol, 3-bromo-2-hydroxynitrobenzene, pubchem3114, 2-brom-6-nitrophenol, 2-nitro-6-bromophenol, acmc-209bjb | |
| OC1=C(Br)C=CC=C1[N+]([O-])=O | |
| 218.01 | |
| 218.01 | |
| Crystalline Powder |
Safety and Handling
TSCA : No