missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog No. BTB04900EA
Change view
Click to view available options
Quantity:
10g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB04900EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB04900EA Supplier Thermo Scientific Chemicals Supplier No. BTB04900EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 113170-71-1
Molecular Formula C7H4BrF3N2O2
Molecular Weight (g/mol) 285.02
MDL Number MFCD00042151
InChI Key ZUZMWPRSGJTLHW-UHFFFAOYSA-N
Synonym 2-bromo-6-nitro-4-trifluoromethyl aniline, 4-amino-3-bromo-5-nitrobenzotrifluoride, 6-bromo-2-nitro-4-trifluoromethyl phenylamine, pubchem2809, 2-bromo-6-nitro-4-trifluoromethyl-phenylamine, 2-bromo-6-nitro-4-trifluoromethyl phenylamine, benzenamine,2-bromo-6-nitro-4-trifluoromethyl, benzenamine, 2-bromo-6-nitro-4-trifluoromethyl
PubChem CID 145492
IUPAC Name 2-bromo-6-nitro-4-(trifluoromethyl)aniline
SMILES NC1=C(Br)C=C(C=C1[N+]([O-])=O)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10g
Formula Weight 285.02
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-nitro-4-(trifluoromethyl)aniline
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.