accessibility menu, dialog, popup

UserName

CAS RN 113170-71-1

IUPAC Name: 2-bromo-6-nitro-4-(trifluoromethyl)aniline
Synonyms: 2-bromo-6-nitro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 285.02
Molecular Formula: C7H4BrF3N2O2
InChi Key: ZUZMWPRSGJTLHW-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(C=C1[N+]([O-])=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)