missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7123345
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40971G 1 g
B40975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40971G Supplier TCI America Supplier No. B40971G

Chemical Identifiers

CAS 90259-31-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD01310788
InChI Key ICXBPDJQFPIBSS-UHFFFAOYSA-N
Synonym 6-bromo-o-toluic acid, 2-bromo-6-methyl-benzoic acid, 6-bromo-2-methylbenzoic acid, benzoic acid, 2-bromo-6-methyl, bromtoluylsaure, bromotoluic acid, pubchem4715, acmc-209r5e, ksc494k9r, 3-bromo-2-carboxytoluene
PubChem CID 2735589
IUPAC Name 2-bromo-6-methylbenzoic acid
SMILES CC1=C(C(O)=O)C(Br)=CC=C1

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 1 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-methylbenzoic Acid