missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-methoxyphenol 98.0+%, TCI America™
Supplier: TCI America B502625G
Specifications
| 2-Bromo-6-methoxyphenol | |
| 64°C | |
| 146°C | |
| MFCD08146628 | |
| 2-bromo-6-methoxy-phenol, phenol, 2-bromo-6-methoxy, bromhydroxyanisol, 6-bromoguaiacol, 3-bromo-2-hydroxyanisole, acmc-1ch0f, phenol,2-bromo-6-methoxy, 2-bromanyl-6-methoxy-phenol, ksc560s4p | |
| COC1=C(C(=CC=C1)Br)O | |
| 203.035 | |
| 203.04 | |
| Crystalline Powder |
| 28165-49-3 | |
| White-Yellow | |
| C7H7BrO2 | |
| 25 g | |
| WEUFQISIJPSTBM-UHFFFAOYSA-N | |
| 2-bromo-6-methoxyphenol | |
| 11019958 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 28165-49-3 | |
| 203.035 | |
| WEUFQISIJPSTBM-UHFFFAOYSA-N | |
| 2-bromo-6-methoxyphenol |
| C7H7BrO2 | |
| MFCD08146628 | |
| 11019958 | |
| COC1=C(C(=CC=C1)Br)O |
Safety and Handling
TSCA : No