missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxyphenol 98.0+%, TCI America™

Catalog No. B50261G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50261G 1 g
B50265G 5 g
B502625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B50261G Supplier TCI America Supplier No. B50261G

Chemical Identifiers

CAS 28165-49-3
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.035
MDL Number MFCD08146628
InChI Key WEUFQISIJPSTBM-UHFFFAOYSA-N
PubChem CID 11019958
IUPAC Name 2-bromo-6-methoxyphenol
SMILES COC1=C(C(=CC=C1)Br)O

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 146°C
Quantity 1 g
Synonym 2-bromo-6-methoxy-phenol, phenol, 2-bromo-6-methoxy, bromhydroxyanisol, 6-bromoguaiacol, 3-bromo-2-hydroxyanisole, acmc-1ch0f, phenol,2-bromo-6-methoxy, 2-bromanyl-6-methoxy-phenol, ksc560s4p
Formula Weight 203.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-methoxyphenol