missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxynaphthalene Pharmaceutical Impurity Standard, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014395
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014395 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014395 Supplier Merck Emd Millipore Supplier No. 0750350MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 5111-65-9
Linear Formula BrC10H6 OCH3
MDL Number MFCD00004062
Melting Point 106°C to 109°C
Synonym Naproxen Impurity N
Type Pharmaceutical Impurity Standard
Quantity 50 mg
Formula Weight 237.09
Percent Purity ≥98.0% (GC)