missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluorobenzaldehyde, 98%

Catalog No. AAH6468906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6468906 5 g
AAH6468914 25 g
AAH6468922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6468906 Supplier Thermo Scientific Chemicals Supplier No. H6468906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 360575-28-6 | C7H4BrFO | 203.01 g/mol

It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.

Chemical Identifiers

CAS 360575-28-6
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD03407341
InChI Key PJNILWKRAKKEQM-UHFFFAOYSA-N
Synonym 6-bromo-2-fluorobenzaldehyde, 2-bromo-6-fluorobenzenecarbaldehyde, 2-bromo-6-fluoro-benzaldehyde, 2-fluoro-6-bromobenzaldehyde, 6-fluoro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-6-fluoro, pubchem1419, acmc-1afpf, ksc497k8t
PubChem CID 22473977
IUPAC Name 2-bromo-6-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Br)C=O)F

Specifications

Melting Point 43°C to 47°C
Density 1.70 g/mL
Boiling Point 100°C to 102°C (8 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2573976
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 203.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-fluorobenzaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only