missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-chlorobenzoic acid, 98%

Catalog No. p-7031259
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3226903 1 g
AAH3226906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3226903 Supplier Thermo Scientific Chemicals Supplier No. H3226903

CAS: 93224-85-2 | C7H4BrClO2 | 235.461 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93224-85-2
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00672929
InChI Key URGXUQODOUMRFP-UHFFFAOYSA-N
Synonym 6-bromo-2-chlorobenzoic acid, 2-chloro-6-bromobenzoic acid, benzoic acid, 2-bromo-6-chloro, 2-bromo-6-chloro-benzoic acid, buttpark 51\01-28, akos msc-0781, 2-bromo-6-chlorobenzoicacid, pubchem4048, acmc-209rkm, 2-chloro-6-bromobenzoic
PubChem CID 33125
IUPAC Name 2-bromo-6-chlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Br)C(=O)O)Cl

Specifications

Melting Point 148°C to 150°C
Quantity 1 g
Formula Weight 235.47
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-chlorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only