missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-chlorobenzaldehyde, 98%

Catalog No. AAH6460503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6460503 1 g
AAH6460506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6460503 Supplier Thermo Scientific Chemicals Supplier No. H6460503

CAS: 64622-16-8 | C7H4BrClO | 219.462 g/mol

2-Bromo-6-chlorobenzaldehyde plays an important role as a linker, which provides higher selectivity and reactivity in the Buchwald C-N bond forming reaction in order to prepare Bruton's tyrosine kinase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-6-chlorobenzaldehyde plays an important role as a linker, which provides higher selectivity and reactivity in the Buchwald C-N bond forming reaction in order to prepare Bruton′s tyrosine kinase inhibitor.

Solubility
Slightly soluble in water. 

Notes
Store in a cool place. Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 64622-16-8
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
MDL Number MFCD00266792
InChI Key NUGMENVSVAURGO-UHFFFAOYSA-N
Synonym 2-chloro-6-bromobenzaldehyde, benzaldehyde, 2-bromo-6-chloro, 2-bromo-6-chloro-benzaldehyde
PubChem CID 13524042
IUPAC Name 2-bromo-6-chlorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Br)C=O)Cl

Specifications

Melting Point 81°C to 86°C
Quantity 1 g
Beilstein 2613087
Solubility Information Slightly soluble in water.
Formula Weight 219.46
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-chlorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only