missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-chloro-4-fluoroaniline 97.0+%, TCI America™

Catalog No. B36815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36815G 5 g
B368125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36815G Supplier TCI America Supplier No. B36815G
Only null left

Chemical Identifiers

CAS 201849-14-1
Molecular Formula C6H4BrClFN
Molecular Weight (g/mol) 224.46
MDL Number MFCD00051707
InChI Key LIBGMUMMWYKJSC-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-6-chloro-4-fluoro, pubchem2922, acmc-209f6h, attercop-chm at130703, 2-bromo-4-fluoro-6-chloroaniline, 2-bromo-6-chloro-4-fluoro-aniline, 2-bromo-6-chloro-4-fluorophenylamine, 6-bromo-2-chloro-4-fluorophenylamine, 2-bromo-6-chloro-4-fluoro-phenylamine, 2-bromo-6-chloro-4-fluoroaniline
PubChem CID 614401
IUPAC Name 2-bromo-6-chloro-4-fluoroaniline
SMILES NC1=C(Cl)C=C(F)C=C1Br

Specifications

Melting Point 57°C
Color Red-White
Quantity 5 g
Formula Weight 224.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-chloro-4-fluoroaniline