missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. B45125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45125G 5 g
B45121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45125G Supplier TCI America Supplier No. B45125G

Chemical Identifiers

CAS 49669-14-9
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD12406888
InChI Key ZRXWMMNBTGBCIL-UHFFFAOYSA-N
Synonym 2-bromo-6-2-methyl-1,3-dioxolan-2-yl pyridine, 2-bromo-6-2-methyl-1,3-dioxolan-2-yl-pyridine, pyridine, 2-bromo-6-2-methyl-1,3-dioxolan-2-yl, pubchem16082, ksc496i5p, 2-bromo-6-2-methyl-1,3-dioxolane-2-yl pyridine, 2-bromanyl-6-2-methyl-1,3-dioxolan-2-yl pyridine, 2-bromo-6-2-methyl-1,3 dioxolan-2-yl-pyridine, 6-bromo-2-2'-methyl-1',3'-dioxolan-2'-yl-pyridine
PubChem CID 12164282
IUPAC Name 2-bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine
SMILES CC1(OCCO1)C2=NC(=CC=C2)Br

Specifications

Melting Point 45°C
Color Yellow
Quantity 5 g
Formula Weight 244.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine