accessibility menu, dialog, popup

UserName

H3COONBr

CAS RN 49669-14-9

IUPAC Name: 2-bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine
Synonyms: 2-bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: ZRXWMMNBTGBCIL-UHFFFAOYSA-N
SMILES: CC1(OCCO1)C1=NC(Br)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results