missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole, Thermo Scientific™
Chemical Identifiers
| CAS | 37461-61-3 |
|---|---|
| Molecular Formula | C3BrF3N2S |
| Molecular Weight (g/mol) | 233.01 |
| MDL Number | MFCD00015535 |
| InChI Key | LGTVLLPQCMJOGT-UHFFFAOYSA-N |
| Synonym | 2-bromo-5-trifluoromethyl-1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4 thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4-thiadiazol, 5-bromo-2-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazole,2-bromo-5-trifluoromethyl, 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl, 2-bromanyl-5-trifluoromethyl-1,3,4-thiadiazole |
| PubChem CID | 12874685 |
| IUPAC Name | 2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole |
| SMILES | FC(F)(F)C1=NN=C(Br)S1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC72310DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 37461-61-3 | |
| 233.01 | |
| LGTVLLPQCMJOGT-UHFFFAOYSA-N | |
| 12874685 | |
| FC(F)(F)C1=NN=C(Br)S1 |
| C3BrF3N2S | |
| MFCD00015535 | |
| 2-bromo-5-trifluoromethyl-1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4 thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4-thiadiazol, 5-bromo-2-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazole,2-bromo-5-trifluoromethyl, 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl, 2-bromanyl-5-trifluoromethyl-1,3,4-thiadiazole | |
| 2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole |
Specifications
| 37461-61-3 | |
| MFCD00015535 | |
| LGTVLLPQCMJOGT-UHFFFAOYSA-N | |
| 2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole | |
| 12874685 | |
| 97% |
| C3BrF3N2S | |
| 2-bromo-5-trifluoromethyl-1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4 thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4-thiadiazol, 5-bromo-2-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazole,2-bromo-5-trifluoromethyl, 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl, 2-bromanyl-5-trifluoromethyl-1,3,4-thiadiazole | |
| FC(F)(F)C1=NN=C(Br)S1 | |
| 233.01 | |
| 233.01 | |
| 2-bromo-5-(trifluoromethyl)-1, 3, 4-thiadiazole |
Safety and Handling
GHS Signal Word: Warning