missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-phenylthiophene 97.0+%, TCI America™
$197.59 - $592.76
Chemical Identifiers
| CAS | 29488-24-2 |
|---|---|
| Molecular Formula | C10H7BrS |
| Molecular Weight (g/mol) | 239.13 |
| MDL Number | MFCD00159575 |
| InChI Key | RVCTZBRMQZWVDA-UHFFFAOYSA-N |
| PubChem CID | 2776313 |
| IUPAC Name | 2-bromo-5-phenylthiophene |
| SMILES | C1=CC=C(C=C1)C2=CC=C(S2)Br |
Chemical Identifiers
| 29488-24-2 | |
| 239.13 | |
| RVCTZBRMQZWVDA-UHFFFAOYSA-N | |
| 2-bromo-5-phenylthiophene |
| C10H7BrS | |
| MFCD00159575 | |
| 2776313 | |
| C1=CC=C(C=C1)C2=CC=C(S2)Br |
Specifications
| 29488-24-2 | |
| White-Yellow | |
| MFCD00159575 | |
| thiophene,2-bromo-5-phenyl, 2-bromo-5-phenyl-thiophene, 2-phenyl-5-bromothiophene, thiophene, 2-bromo-5-phenyl, 5-bromo-2-phenylthiophene, acmc-209h9c, 2-bromo-5-phenyl thiophene | |
| C1=CC=C(C=C1)C2=CC=C(S2)Br | |
| 239.13 | |
| 239.13 | |
| Crystalline Powder |
| 85°C | |
| C10H7BrS | |
| 1 g | |
| RVCTZBRMQZWVDA-UHFFFAOYSA-N | |
| 2-bromo-5-phenylthiophene | |
| 2776313 | |
| ≥97.0% (GC) | |
| 2-Bromo-5-phenylthiophene |
Safety and Handling
TSCA : No